missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Nitrobenzyl Alcohol 98.0+%, TCI America™
$61.51 - $111.84
Chemical Identifiers
| CAS | 619-73-8 |
|---|---|
| Molecular Formula | C7H7NO3 |
| Molecular Weight (g/mol) | 153.14 |
| MDL Number | MFCD00007376 |
| InChI Key | JKTYGPATCNUWKN-UHFFFAOYSA-N |
| Synonym | 4-nitrobenzyl alcohol, 4-nitrophenyl methanol, p-nitrobenzyl alcohol, benzenemethanol, 4-nitro, 4-nitrobenzenemethanol, p-hydroxymethyl nitrobenzene, benzyl alcohol, p-nitro, paranitrobenzyl alcohol, 4-nitrobenzylalcohol, unii-86btj68y9m |
| PubChem CID | 69275 |
| ChEBI | CHEBI:41214 |
| IUPAC Name | (4-nitrophenyl)methanol |
| SMILES | OCC1=CC=C(C=C1)[N+]([O-])=O |
Chemical Identifiers
| 619-73-8 | |
| 153.14 | |
| JKTYGPATCNUWKN-UHFFFAOYSA-N | |
| 69275 | |
| (4-nitrophenyl)methanol |
| C7H7NO3 | |
| MFCD00007376 | |
| 4-nitrobenzyl alcohol, 4-nitrophenyl methanol, p-nitrobenzyl alcohol, benzenemethanol, 4-nitro, 4-nitrobenzenemethanol, p-hydroxymethyl nitrobenzene, benzyl alcohol, p-nitro, paranitrobenzyl alcohol, 4-nitrobenzylalcohol, unii-86btj68y9m | |
| CHEBI:41214 | |
| OCC1=CC=C(C=C1)[N+]([O-])=O |
Specifications
| 619-73-8 | |
| 185°C | |
| MFCD00007376 | |
| 4-nitrobenzyl alcohol, 4-nitrophenyl methanol, p-nitrobenzyl alcohol, benzenemethanol, 4-nitro, 4-nitrobenzenemethanol, p-hydroxymethyl nitrobenzene, benzyl alcohol, p-nitro, paranitrobenzyl alcohol, 4-nitrobenzylalcohol, unii-86btj68y9m | |
| OCC1=CC=C(C=C1)[N+]([O-])=O | |
| 153.14 | |
| CHEBI:41214 | |
| ≥98.0% (GC) | |
| 4-Nitrobenzyl Alcohol |
| 95°C | |
| C7H7NO3 | |
| 25 g | |
| JKTYGPATCNUWKN-UHFFFAOYSA-N | |
| (4-nitrophenyl)methanol | |
| 69275 | |
| 153.14 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (3)-3797
RTECSNumber : DP0657100
TSCA : Yes