missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Nitrobenzyl Alcohol 98.0+%, TCI America™
Supplier: TCI America N018025G
Specifications
| 4-Nitrobenzyl Alcohol | |
| 95°C | |
| C7H7NO3 | |
| 25 g | |
| JKTYGPATCNUWKN-UHFFFAOYSA-N | |
| (4-nitrophenyl)methanol | |
| 69275 | |
| 153.14 | |
| Crystalline Powder |
| 619-73-8 | |
| 185°C | |
| MFCD00007376 | |
| 4-nitrobenzyl alcohol, 4-nitrophenyl methanol, p-nitrobenzyl alcohol, benzenemethanol, 4-nitro, 4-nitrobenzenemethanol, p-hydroxymethyl nitrobenzene, benzyl alcohol, p-nitro, paranitrobenzyl alcohol, 4-nitrobenzylalcohol, unii-86btj68y9m | |
| OCC1=CC=C(C=C1)[N+]([O-])=O | |
| 153.14 | |
| CHEBI:41214 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 619-73-8 | |
| 153.14 | |
| JKTYGPATCNUWKN-UHFFFAOYSA-N | |
| 69275 | |
| (4-nitrophenyl)methanol |
| C7H7NO3 | |
| MFCD00007376 | |
| 4-nitrobenzyl alcohol, 4-nitrophenyl methanol, p-nitrobenzyl alcohol, benzenemethanol, 4-nitro, 4-nitrobenzenemethanol, p-hydroxymethyl nitrobenzene, benzyl alcohol, p-nitro, paranitrobenzyl alcohol, 4-nitrobenzylalcohol, unii-86btj68y9m | |
| CHEBI:41214 | |
| OCC1=CC=C(C=C1)[N+]([O-])=O |
Safety and Handling
EINECSNumber : (3)-3797
RTECSNumber : DP0657100
TSCA : Yes