missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzyl Acetate 98.0+%, TCI America™

Catalog No. A003925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A003925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A003925G Supplier TCI America Supplier No. A003925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 619-90-9
Molecular Formula C9H9NO4
Molecular Weight (g/mol) 195.174
MDL Number MFCD00024781
InChI Key QNXQLPUEZYZYFC-UHFFFAOYSA-N
Synonym Acetic Acid 4-Nitrobenzyl Ester
PubChem CID 12094
IUPAC Name (4-nitrophenyl)methyl acetate
SMILES CC(=O)OCC1=CC=C(C=C1)[N+](=O)[O-]

Specifications

Melting Point 78°C
Color Yellow
Quantity 25 g
Formula Weight 195.17
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrobenzyl Acetate
RTECSNumber DP0660000
TSCA Yes