accessibility menu, dialog, popup

UserName

CAS RN 619-90-9

IUPAC Name: (4-nitrophenyl)methyl acetate
Synonyms: acetic acid, nitrobenzyl ester
Molecular Weight (g/mol): 195.17
Molecular Formula: C9H9NO4
InChi Key: QNXQLPUEZYZYFC-UHFFFAOYSA-N
SMILES: CC(=O)OCC1=CC=C(C=C1)[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results