missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Nitrobenzotrifluoride 98.0+%, TCI America™
Supplier: TCI America N090225G
Specifications
| 4-Nitrobenzotrifluoride | |
| 42°C | |
| C7H4F3NO2 | |
| 25 g | |
| 4-nitrobenzotrifluoride, 1-nitro-4-trifluoromethyl benzene, p-nitrobenzotrifluoride, benzene, 1-nitro-4-trifluoromethyl, 4-nitro-alpha,alpha,alpha-trifluorotoluene, 4-trifluoromethyl nitrobenzene, 4-nitro-a,a,a-trifluorotoluene, p-nitro-trifluoromethylphenol, p-trifluoromethylnitrobenzene, 4-nitro-.alpha.,.alpha.,.alpha.-trifluorotoluene | |
| C1=CC(=CC=C1C(F)(F)F)[N+](=O)[O-] | |
| 191.109 | |
| 191.11 | |
| Crystalline Lumps |
| 402-54-0 | |
| 82°C | |
| MFCD00007358 | |
| 3431 | |
| XKYLCLMYQDFGKO-UHFFFAOYSA-N | |
| 1-nitro-4-(trifluoromethyl)benzene | |
| 9821 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 402-54-0 | |
| 191.109 | |
| XKYLCLMYQDFGKO-UHFFFAOYSA-N | |
| 9821 | |
| C1=CC(=CC=C1C(F)(F)F)[N+](=O)[O-] |
| C7H4F3NO2 | |
| MFCD00007358 | |
| 4-nitrobenzotrifluoride, 1-nitro-4-trifluoromethyl benzene, p-nitrobenzotrifluoride, benzene, 1-nitro-4-trifluoromethyl, 4-nitro-alpha,alpha,alpha-trifluorotoluene, 4-trifluoromethyl nitrobenzene, 4-nitro-a,a,a-trifluorotoluene, p-nitro-trifluoromethylphenol, p-trifluoromethylnitrobenzene, 4-nitro-.alpha.,.alpha.,.alpha.-trifluorotoluene | |
| 1-nitro-4-(trifluoromethyl)benzene |
Safety and Handling
TSCA : No