missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzotrifluoride 98.0+%, TCI America™

Catalog No. p-7134020
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N09025G 5 g
N090225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N09025G Supplier TCI America Supplier No. N09025G
Only null left

Chemical Identifiers

CAS 402-54-0
Molecular Formula C7H4F3NO2
Molecular Weight (g/mol) 191.109
MDL Number MFCD00007358
InChI Key XKYLCLMYQDFGKO-UHFFFAOYSA-N
Synonym 4-nitrobenzotrifluoride, 1-nitro-4-trifluoromethyl benzene, p-nitrobenzotrifluoride, benzene, 1-nitro-4-trifluoromethyl, 4-nitro-alpha,alpha,alpha-trifluorotoluene, 4-trifluoromethyl nitrobenzene, 4-nitro-a,a,a-trifluorotoluene, p-nitro-trifluoromethylphenol, p-trifluoromethylnitrobenzene, 4-nitro-.alpha.,.alpha.,.alpha.-trifluorotoluene
PubChem CID 9821
IUPAC Name 1-nitro-4-(trifluoromethyl)benzene
SMILES C1=CC(=CC=C1C(F)(F)F)[N+](=O)[O-]

Specifications

Melting Point 42°C
Boiling Point 82°C
Quantity 5 g
UN Number 3431
Formula Weight 191.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Nitrobenzotrifluoride