missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzenediazonium tetrafluoroborate, 97%

Catalog No. p-7036866
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5531503 1 g
AAH5531506 5 g
AAH5531514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH5531503 Supplier Thermo Scientific Chemicals Supplier No. H5531503
Only null left

CAS: 456-27-9 | C6H4BF4N3O2 | 236.92 g/mol

4-Nitrobenzenediazonium tetrafluoroborate is widely used as a standard reagent in organic synthesis. It is used as a dye for enterochromaffin cells. It is used for the spectrophotometric determination of common aromatic diisocyanates [toluene-2,4-diyl diisocyanate and 4,4'-methylenebis(phenyl isocyanate)] and gallic acid. It is also used for tissue staining and determination of trace phenols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrobenzenediazonium tetrafluoroborate is widely used as a standard reagent in organic synthesis. It is used as a dye for enterochromaffin cells. It is used for the spectrophotometric determination of common aromatic diisocyanates [toluene-2,4-diyl diisocyanate and 4,4′-methylenebis(phenyl isocyanate)] and gallic acid. It is also used for tissue staining and determination of trace phenols.

Solubility
Soluble in acetonitrile and dimethyl sulfoxide.

Notes
Moisture sensitive. Store in a cool place. Keep the container tightly closed. Incompatible with strong bases, strong oxidizing agents and moisture.

Chemical Identifiers

CAS 456-27-9
Molecular Formula C6H4BF4N3O2
Molecular Weight (g/mol) 236.92
MDL Number MFCD00012005
InChI Key AFULQCYCQAHYIP-UHFFFAOYSA-N
Synonym 4-nitrobenzenediazonium tetrafluoroborate, ccris 3641, unii-2uxo615r4o, p-nitrobenzenediazonium tetrafluoroborate, benzenediazonium, 4-nitro-, tetrafluoroborate 1-, p-nitrophenyldiazonium fluoborate, p-nitrobenzenediazonium fluoborate, 4-nitrobenzenediazonium fluoborate, p-nitrobenzenediazonium fluoroborate, 4-nitrobenzenediazonium fluoroborate
PubChem CID 68006
IUPAC Name 4-nitrobenzenediazonium;tetrafluoroborate
SMILES F[B-](F)(F)F.[O-][N+](=O)C1=CC=C(C=C1)[N+]#N

Specifications

Melting Point 144°C to 148°C
Quantity 1 g
UN Number UN2923
Beilstein 3599869
Solubility Information Soluble in acetonitrile and dimethyl sulfoxide.
Formula Weight 236.92
Percent Purity 97%
Chemical Name or Material 4-Nitrobenzenediazonium tetrafluoroborate
Hazard Category H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H314-H318-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
EINECSNumber 207-261-1
RTECSNumber CZ1723000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only