missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Nitrobenzaldehyde 98.0+%, TCI America™
Supplier: TCI America N0559100G
Specifications
| 4-Nitrobenzaldehyde | |
| 105°C | |
| MFCD00007346 | |
| p-nitrobenzaldehyde, benzaldehyde, 4-nitro, p-formylnitrobenzene, benzaldehyde, p-nitro, 4-nitrobenzaldehydde, 4-nitro-benzaldehyde, 4-formylnitrobenzene, p-nitro benzaldehyde, 4-nitro benzaldehyde, ccris 1675 | |
| [O-][N+](=O)C1=CC=C(C=O)C=C1 | |
| 151.12 | |
| CHEBI:66926 | |
| ≥98.0% (GC) |
| 555-16-8 | |
| C7H5NO3 | |
| 100 g | |
| BXRFQSNOROATLV-UHFFFAOYSA-N | |
| 4-nitrobenzaldehyde | |
| 541 | |
| 151.12 | |
| Crystalline Powder |
Chemical Identifiers
| 555-16-8 | |
| 151.12 | |
| BXRFQSNOROATLV-UHFFFAOYSA-N | |
| 541 | |
| 4-nitrobenzaldehyde |
| C7H5NO3 | |
| MFCD00007346 | |
| p-nitrobenzaldehyde, benzaldehyde, 4-nitro, p-formylnitrobenzene, benzaldehyde, p-nitro, 4-nitrobenzaldehydde, 4-nitro-benzaldehyde, 4-formylnitrobenzene, p-nitro benzaldehyde, 4-nitro benzaldehyde, ccris 1675 | |
| CHEBI:66926 | |
| [O-][N+](=O)C1=CC=C(C=O)C=C1 |
Safety and Handling
EINECSNumber : (3)-1181
RTECSNumber : CU7350000
TSCA : Yes