missing translation for 'onlineSavingsMsg'
Learn More

4-Nitro-p-terphenyl 95.0+%, TCI America™

Catalog No. N06101G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N06101G 1 g
N06105G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N06101G Supplier TCI America Supplier No. N06101G

Chemical Identifiers

CAS 10355-53-0
Molecular Formula C18H13NO2
Molecular Weight (g/mol) 275.307
MDL Number MFCD00044846
InChI Key IMMGNSJGTWWGJB-UHFFFAOYSA-N
Synonym 4-nitro-p-terphenyl, p-terphenyl, 4-nitro, 1-4-nitrophenyl-4-phenylbenzene, 4-nitro-1,1':4',1-terphenyl, 1,1':4',1-terphenyl, 4-nitro, 4-nitro-1-4-phenylphenyl benzene, 4-nitro-1,1':4':1'-terphenyl, 4-nitro-para-terphenyl, 3-05-00-02299 beilstein handbook reference, 1,1-terphenyl, 4-nitro
PubChem CID 25196
IUPAC Name 1-(4-nitrophenyl)-4-phenylbenzene
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)C3=CC=C(C=C3)[N+](=O)[O-]

Specifications

Melting Point 214°C
Color Yellow
Quantity 1 g
Formula Weight 275.31
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitro-p-terphenyl
RTECSNumber WZ6525000
TSCA No