accessibility menu, dialog, popup

UserName

CAS RN 10355-53-0

IUPAC Name: 4-(4-nitrophenyl)-1,1'-biphenyl
Synonyms: 4-(4-nitrophenyl)-1,1'-biphenyl
Molecular Weight (g/mol): 275.31
Molecular Formula: C18H13NO2
InChi Key: IMMGNSJGTWWGJB-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(C=C1)C1=CC=C(C=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results