missing translation for 'onlineSavingsMsg'
Learn More

4-Nitro-3-(trifluoromethyl)phenol 99.0+%, TCI America™

Catalog No. p-7133724
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
N02795G 5 g
N027925G 25 g
2 options

Catalog No. N02795G

Supplier: TCI America N02795G

Only null left

Chemical Identifiers

CAS 88-30-2
Molecular Formula C7H4F3NO3
Molecular Weight (g/mol) 207.108
MDL Number MFCD00014714
InChI Key ZEFMBAFMCSYJOO-UHFFFAOYSA-N
Synonym 4-nitro-3-trifluoromethyl phenol, lamprecide, lamprecid, 5-hydroxy-2-nitrobenzotrifluoride, dowlap f, 3-trifluoromethyl-4-nitrophenol, phenol, 4-nitro-3-trifluoromethyl, usaf ma-6, caswell no. 890, tfm piscicide
PubChem CID 6931
IUPAC Name 4-nitro-3-(trifluoromethyl)phenol
SMILES C1=CC(=C(C=C1O)C(F)(F)F)[N+](=O)[O-]

Specifications

Melting Point 80°C
Color Yellow
Quantity 5 g
UN Number 2811
Formula Weight 207.11
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitro-3-(trifluoromethyl)phenol
RTECSNumber GP3520000
TSCA No