accessibility menu, dialog, popup

UserName

CAS RN 88-30-2

IUPAC Name: 4-nitro-3-(trifluoromethyl)phenol
Synonyms: tfm (piscicide)
Molecular Weight (g/mol): 207.11
Molecular Formula: C7H4F3NO3
InChi Key: ZEFMBAFMCSYJOO-UHFFFAOYSA-N
SMILES: OC1=CC=C(C(=C1)C(F)(F)F)[N+]([O-])=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)