missing translation for 'onlineSavingsMsg'
Learn More

4-Nitro-3-(trifluoromethyl)phenol, 97%

CAS: 88-30-2 | C7H4F3NO3 | 207.108 g/mol

Supplier:  Thermo Scientific Chemicals L0822806

 View more versions of this product

Catalog No. AAL0822806

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Nitro-3-(trifluoromethyl)phenol
74°C to 77°C
C7H4F3NO3
5 g
2053698
4-nitro-3-trifluoromethyl phenol, lamprecide, lamprecid, 5-hydroxy-2-nitrobenzotrifluoride, dowlap f, 3-trifluoromethyl-4-nitrophenol, phenol, 4-nitro-3-trifluoromethyl, usaf ma-6, caswell no. 890, tfm piscicide
C1=CC(=C(C=C1O)C(F)(F)F)[N+](=O)[O-]
207.108
207.11
88-30-2
135°C to 138°C (0.01 mmHg)
MFCD00014714
UN2811
14,9253
ZEFMBAFMCSYJOO-UHFFFAOYSA-N
4-nitro-3-(trifluoromethyl)phenol
6931
97%

Chemical Identifiers

88-30-2
207.108
ZEFMBAFMCSYJOO-UHFFFAOYSA-N
6931
C1=CC(=C(C=C1O)C(F)(F)F)[N+](=O)[O-]
C7H4F3NO3
MFCD00014714
4-nitro-3-trifluoromethyl phenol, lamprecide, lamprecid, 5-hydroxy-2-nitrobenzotrifluoride, dowlap f, 3-trifluoromethyl-4-nitrophenol, phenol, 4-nitro-3-trifluoromethyl, usaf ma-6, caswell no. 890, tfm piscicide
4-nitro-3-(trifluoromethyl)phenol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 201-818-2

RTECSNumber : GP3520000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only