Learn More
4-Nitro-3-(trifluoromethyl)aniline, 98%
CAS: 393-11-3 | C7H5F3N2O2 | 206.12 g/mol
$65.84 - $199.48
Chemical Identifiers
| CAS | 393-11-3 |
|---|---|
| Molecular Formula | C7H5F3N2O2 |
| Molecular Weight (g/mol) | 206.12 |
| MDL Number | MFCD00014717 |
| InChI Key | UTKUVRNVYFTEHF-UHFFFAOYSA-N |
| Synonym | 4-nitro-3-trifluoromethyl aniline, 5-amino-2-nitrobenzotrifluoride, 3-amino-6-nitrobenzotrifluoride, benzenamine, 4-nitro-3-trifluoromethyl, 3-trifluoromethyl-4-nitroaniline, 2-nitro-5-amino benzotrifluoride, 3-trifluoromethyl-4-nitroanilide, flu-1, 3-amino-6-nitro benzotrifluoride |
| PubChem CID | 94955 |
| IUPAC Name | 4-nitro-3-(trifluoromethyl)aniline |
| SMILES | NC1=CC=C(C(=C1)C(F)(F)F)[N+]([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1379803
|
Thermo Scientific Chemicals
A1379803 |
1 g |
N/A
|
|
|||||
|
AAA1379806
|
Thermo Scientific Chemicals
A1379806 |
5 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 393-11-3 | |
| 206.12 | |
| UTKUVRNVYFTEHF-UHFFFAOYSA-N | |
| 94955 | |
| NC1=CC=C(C(=C1)C(F)(F)F)[N+]([O-])=O |
| C7H5F3N2O2 | |
| MFCD00014717 | |
| 4-nitro-3-trifluoromethyl aniline, 5-amino-2-nitrobenzotrifluoride, 3-amino-6-nitrobenzotrifluoride, benzenamine, 4-nitro-3-trifluoromethyl, 3-trifluoromethyl-4-nitroaniline, 2-nitro-5-amino benzotrifluoride, 3-trifluoromethyl-4-nitroanilide, flu-1, 3-amino-6-nitro benzotrifluoride | |
| 4-nitro-3-(trifluoromethyl)aniline |
Specifications
| 393-11-3 | |
| C7H5F3N2O2 | |
| 1 g | |
| 2650702 | |
| UTKUVRNVYFTEHF-UHFFFAOYSA-N | |
| 4-nitro-3-(trifluoromethyl)aniline | |
| 94955 | |
| 98% |
| 125°C to 128°C | |
| MFCD00014717 | |
| UN2811 | |
| 4-nitro-3-trifluoromethyl aniline, 5-amino-2-nitrobenzotrifluoride, 3-amino-6-nitrobenzotrifluoride, benzenamine, 4-nitro-3-trifluoromethyl, 3-trifluoromethyl-4-nitroaniline, 2-nitro-5-amino benzotrifluoride, 3-trifluoromethyl-4-nitroanilide, flu-1, 3-amino-6-nitro benzotrifluoride | |
| NC1=CC=C(C(=C1)C(F)(F)F)[N+]([O-])=O | |
| 206.12 | |
| 206.13 | |
| 4-Nitro-3-(trifluoromethyl)aniline |
Safety and Handling
GHS H Statement
H311-H302-H332-H315-H319-H335
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
H302+H312+H332
DOTInformation : Hazard Class: 6.1; Packaging Group: II
EINECSNumber : 206-884-6
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only