missing translation for 'onlineSavingsMsg'
Learn More

4-Nitro-3-(trifluoromethyl)aniline, 98%

CAS: 393-11-3 | C7H5F3N2O2 | 206.12 g/mol

Supplier:  Thermo Scientific Chemicals A1379806

 View more versions of this product

Catalog No. AAA1379806


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Nitro-3-(trifluoromethyl)aniline
125°C to 128°C
MFCD00014717
UN2811
4-nitro-3-trifluoromethyl aniline, 5-amino-2-nitrobenzotrifluoride, 3-amino-6-nitrobenzotrifluoride, benzenamine, 4-nitro-3-trifluoromethyl, 3-trifluoromethyl-4-nitroaniline, 2-nitro-5-amino benzotrifluoride, 3-trifluoromethyl-4-nitroanilide, flu-1, 3-amino-6-nitro benzotrifluoride
NC1=CC=C(C(=C1)C(F)(F)F)[N+]([O-])=O
206.12
206.13
393-11-3
C7H5F3N2O2
5 g
2650702
UTKUVRNVYFTEHF-UHFFFAOYSA-N
4-nitro-3-(trifluoromethyl)aniline
94955
98%

Chemical Identifiers

393-11-3
206.12
UTKUVRNVYFTEHF-UHFFFAOYSA-N
94955
NC1=CC=C(C(=C1)C(F)(F)F)[N+]([O-])=O
C7H5F3N2O2
MFCD00014717
4-nitro-3-trifluoromethyl aniline, 5-amino-2-nitrobenzotrifluoride, 3-amino-6-nitrobenzotrifluoride, benzenamine, 4-nitro-3-trifluoromethyl, 3-trifluoromethyl-4-nitroaniline, 2-nitro-5-amino benzotrifluoride, 3-trifluoromethyl-4-nitroanilide, flu-1, 3-amino-6-nitro benzotrifluoride
4-nitro-3-(trifluoromethyl)aniline
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H311-H302-H332-H315-H319-H335
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c

H302+H312+H332

DOTInformation : Hazard Class: 6.1; Packaging Group: II

EINECSNumber : 206-884-6

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only