missing translation for 'onlineSavingsMsg'
Learn More

4'-Nitro-3'-(trifluoromethyl)acetanilide 98.0+%, TCI America™

Catalog No. p-7133867
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N05485G 5 g
N054825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N05485G Supplier TCI America Supplier No. N05485G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 393-12-4
Molecular Formula C9H7F3N2O3
Molecular Weight (g/mol) 248.161
MDL Number MFCD00017994
InChI Key MIHJCLQINRFOLX-UHFFFAOYSA-N
PubChem CID 246769
IUPAC Name N-[4-nitro-3-(trifluoromethyl)phenyl]acetamide
SMILES CC(=O)NC1=CC(=C(C=C1)[N+](=O)[O-])C(F)(F)F

Specifications

Melting Point 123°C
Quantity 5 g
Formula Weight 248.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Nitro-3′-(trifluoromethyl)acetanilide