accessibility menu, dialog, popup

UserName

CAS RN 393-12-4

IUPAC Name: N-[4-nitro-3-(trifluoromethyl)phenyl]acetamide
Synonyms: N-[4-nitro-3-(trifluoromethyl)phenyl]acetamide
Molecular Weight (g/mol): 248.16
Molecular Formula: C9H7F3N2O3
InChi Key: MIHJCLQINRFOLX-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=C(C(=C1)C(F)(F)F)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results