missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Nitro-2,1,3-benzothiadiazole 98.0+%, TCI America™
Supplier: TCI America N078625G
Specifications
| 4-Nitro-2,1,3-benzothiadiazole | |
| 109°C | |
| MFCD00068050 | |
| 4-nitrobenzo c 1,2,5 thiadiazole, 4-nitropiazthiole, 2,1,3-benzothiadiazole, 4-nitro, 4-nitro-1,2,3-benzothiadiazole, 4-nitro-benzo 1,2,5 thiadiazole, 4-nitro-2??,1,3-benzothiadiazole, 4-nitrobenzo c 1,2,5-thiadiazole, d0v8fg, acmc-1b59l, 4-bromo-2,8-dimethylphenol | |
| C1=CC2=NSN=C2C(=C1)[N+](=O)[O-] | |
| 181.17 | |
| 181.17 | |
| Crystalline Powder |
| 6583-06-8 | |
| C6H3N3O2S | |
| 25 g | |
| IWQKAMJGVIHECB-UHFFFAOYSA-N | |
| 4-nitro-2,1,3-benzothiadiazole | |
| 81062 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 6583-06-8 | |
| 181.17 | |
| IWQKAMJGVIHECB-UHFFFAOYSA-N | |
| 81062 | |
| C1=CC2=NSN=C2C(=C1)[N+](=O)[O-] |
| C6H3N3O2S | |
| MFCD00068050 | |
| 4-nitrobenzo c 1,2,5 thiadiazole, 4-nitropiazthiole, 2,1,3-benzothiadiazole, 4-nitro, 4-nitro-1,2,3-benzothiadiazole, 4-nitro-benzo 1,2,5 thiadiazole, 4-nitro-2??,1,3-benzothiadiazole, 4-nitrobenzo c 1,2,5-thiadiazole, d0v8fg, acmc-1b59l, 4-bromo-2,8-dimethylphenol | |
| 4-nitro-2,1,3-benzothiadiazole |
Safety and Handling
TSCA : No