CAS RN 6583-06-8
CAS RN 6583-06-8
IUPAC Name:
4-nitro-2,1,3-benzothiadiazole
Synonyms:
4-nitro-2,1,3-benzothiadiazole
Molecular Weight (g/mol):
181.17
Molecular Formula:
C6H3N3O2S
InChi Key:
IWQKAMJGVIHECB-UHFFFAOYSA-N
SMILES:
[O-][N+](=O)C1=CC=CC2=NSN=C12
1
–
1
of
1
results
1
4-Nitro-2,1,3-benzothiadiazole 98.0+%, TCI America™
CAS: 6583-06-8 Molecular Formula: C6H3N3O2S Molecular Weight (g/mol): 181.17 MDL Number: MFCD00068050 InChI Key: IWQKAMJGVIHECB-UHFFFAOYSA-N Synonym: 4-nitrobenzo c 1,2,5 thiadiazole,4-nitropiazthiole,2,1,3-benzothiadiazole, 4-nitro,4-nitro-1,2,3-benzothiadiazole,4-nitro-benzo 1,2,5 thiadiazole,4-nitro-2??,1,3-benzothiadiazole,4-nitrobenzo c 1,2,5-thiadiazole,d0v8fg,acmc-1b59l,4-bromo-2,8-dimethylphenol PubChem CID: 81062 IUPAC Name: 4-nitro-2,1,3-benzothiadiazole SMILES: C1=CC2=NSN=C2C(=C1)[N+](=O)[O-]