missing translation for 'onlineSavingsMsg'
Learn More

4-Nitro-2,1,3-benzothiadiazole 98.0+%, TCI America™

Catalog No. p-7133973
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N07865G 5 g
N078625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N07865G Supplier TCI America Supplier No. N07865G
Only null left

Chemical Identifiers

CAS 6583-06-8
Molecular Formula C6H3N3O2S
Molecular Weight (g/mol) 181.17
MDL Number MFCD00068050
InChI Key IWQKAMJGVIHECB-UHFFFAOYSA-N
Synonym 4-nitrobenzo c 1,2,5 thiadiazole, 4-nitropiazthiole, 2,1,3-benzothiadiazole, 4-nitro, 4-nitro-1,2,3-benzothiadiazole, 4-nitro-benzo 1,2,5 thiadiazole, 4-nitro-2??,1,3-benzothiadiazole, 4-nitrobenzo c 1,2,5-thiadiazole, d0v8fg, acmc-1b59l, 4-bromo-2,8-dimethylphenol
PubChem CID 81062
IUPAC Name 4-nitro-2,1,3-benzothiadiazole
SMILES C1=CC2=NSN=C2C(=C1)[N+](=O)[O-]

Specifications

Melting Point 109°C
Quantity 5 g
Formula Weight 181.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitro-2,1,3-benzothiadiazole