missing translation for 'onlineSavingsMsg'
Learn More

4-n-Propylphenylacetylene, 97%

Catalog No. p-7027972
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2506506 5 g
AAB2506514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2506506 Supplier Thermo Scientific Chemicals Supplier No. B2506506
Only null left

CAS: 62452-73-7 | C11H12 | 144.217 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 62452-73-7
Molecular Formula C11H12
Molecular Weight (g/mol) 144.217
MDL Number MFCD00173886
InChI Key UVFFOABHOIMLNB-UHFFFAOYSA-N
Synonym 4-propylphenylacetylene, 1-eth-1-ynyl-4-propylbenzene, 4-n-propylphenylacetylene, 1-ethynyl-4-propyl-benzene, benzene, 1-ethynyl-4-propyl, pubchem10973, 4-n-propylphenyl ethine, 4-propyl phenylacetylene, 4-ethynyl-1-propylbenzene, 4-propyl-1-ethynylbenzene
PubChem CID 2775133
IUPAC Name 1-ethynyl-4-propylbenzene
SMILES CCCC1=CC=C(C=C1)C#C

Specifications

Density 0.91
Boiling Point 205°C
Flash Point 67°C (153°F)
Refractive Index 1.534
Quantity 5 g
Formula Weight 144.22
Percent Purity 97%
Chemical Name or Material 4-n-Propylphenylacetylene
Hazard Category H227-H315-H319-H335
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only