accessibility menu, dialog, popup

UserName

CAS RN 62452-73-7

IUPAC Name: 1-ethynyl-4-propylbenzene
Synonyms: 1-ethynyl-4-propylbenzene
Molecular Weight (g/mol): 144.22
Molecular Formula: C11H12
InChi Key: UVFFOABHOIMLNB-UHFFFAOYSA-N
SMILES: CCCC1=CC=C(C=C1)C#C
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (5)
  • (2)
  • (5)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)