missing translation for 'onlineSavingsMsg'
Learn More

4-n-Propylphenol, 98%

CAS: 645-56-7 | C9H12O | 136.194 g/mol

$90.92 - $965.62

Chemical Identifiers

CAS 645-56-7
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00002395
InChI Key KLSLBUSXWBJMEC-UHFFFAOYSA-N
Synonym 4-n-propylphenol, p-propylphenol, phenol, 4-propyl, dihydrochavicol, phenol, p-propyl, 1-hydroxy-4-n-propylbenzene, p-n-propylphenol, 4-propyl-phenol, p-hydroxypropylbenzene, hydroxypropylbenzene, p
PubChem CID 12580
ChEBI CHEBI:34434
IUPAC Name 4-propylphenol
SMILES CCCC1=CC=C(C=C1)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Qty  
Catalog Number Mfr. No. Quantity Price Qty  
AAA1534206
View Documents
Thermo Scientific Chemicals
A1534206
5 g
Each for $90.92
Only null left
 
AAA1534214
View Documents
Thermo Scientific Chemicals
A1534214
25 g
Each for $294.81
Only null left
 
AAA1534222
View Documents
Thermo Scientific Chemicals
A1534222
100 g
Each for $965.62
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

645-56-7
136.194
KLSLBUSXWBJMEC-UHFFFAOYSA-N
12580
4-propylphenol
C9H12O
MFCD00002395
4-n-propylphenol, p-propylphenol, phenol, 4-propyl, dihydrochavicol, phenol, p-propyl, 1-hydroxy-4-n-propylbenzene, p-n-propylphenol, 4-propyl-phenol, p-hydroxypropylbenzene, hydroxypropylbenzene, p
CHEBI:34434
CCCC1=CC=C(C=C1)O

Specifications

645-56-7
0.983
106°C (222°F)
1.523
5 g
1363429
KLSLBUSXWBJMEC-UHFFFAOYSA-N
4-propylphenol
12580
136.19
4-n-Propylphenol
20°C to 22°C
232°C
C9H12O
MFCD00002395
UN3145
4-n-propylphenol, p-propylphenol, phenol, 4-propyl, dihydrochavicol, phenol, p-propyl, 1-hydroxy-4-n-propylbenzene, p-n-propylphenol, 4-propyl-phenol, p-hydroxypropylbenzene, hydroxypropylbenzene, p
CCCC1=CC=C(C=C1)O
136.194
CHEBI:34434
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302+H312-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ALKYLPHENOLS, LIQUID, N.O.S.

EINECSNumber : 211-446-2

RTECSNumber : SM8610000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only