missing translation for 'onlineSavingsMsg'
Learn More

4-n-Propylphenol, 98%

CAS: 645-56-7 | C9H12O | 136.194 g/mol

Supplier:  Thermo Scientific Chemicals A1534214

 View more versions of this product

Catalog No. AAA1534214


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-n-Propylphenol
20°C to 22°C
232°C
C9H12O
MFCD00002395
UN3145
4-n-propylphenol, p-propylphenol, phenol, 4-propyl, dihydrochavicol, phenol, p-propyl, 1-hydroxy-4-n-propylbenzene, p-n-propylphenol, 4-propyl-phenol, p-hydroxypropylbenzene, hydroxypropylbenzene, p
CCCC1=CC=C(C=C1)O
136.194
CHEBI:34434
98%
645-56-7
0.983
106°C (222°F)
1.523
25 g
1363429
KLSLBUSXWBJMEC-UHFFFAOYSA-N
4-propylphenol
12580
136.19

Chemical Identifiers

645-56-7
136.194
KLSLBUSXWBJMEC-UHFFFAOYSA-N
12580
4-propylphenol
C9H12O
MFCD00002395
4-n-propylphenol, p-propylphenol, phenol, 4-propyl, dihydrochavicol, phenol, p-propyl, 1-hydroxy-4-n-propylbenzene, p-n-propylphenol, 4-propyl-phenol, p-hydroxypropylbenzene, hydroxypropylbenzene, p
CHEBI:34434
CCCC1=CC=C(C=C1)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302+H312-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ALKYLPHENOLS, LIQUID, N.O.S.

EINECSNumber : 211-446-2

RTECSNumber : SM8610000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only