missing translation for 'onlineSavingsMsg'
Learn More

4-n-Propoxybenzoic acid, 98%

Catalog No. p-7027897
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2495806 5 g
AAB2495814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2495806 Supplier Thermo Scientific Chemicals Supplier No. B2495806

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5438-19-7 | C10H12O3 | 180.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5438-19-7
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.20
MDL Number MFCD00013989
InChI Key GDFUWFOCYZZGQU-UHFFFAOYSA-N
Synonym p-propoxybenzoic acid, 4-n-propoxybenzoic acid, benzoic acid, 4-propoxy, 4-n-propyloxybenzoic acid, 4-propoxy-benzoic acid, benzoic acid, p-propoxy, p-propyloxybenzoic acid, pubchem2681, 4-propoxy benzoic acid, p-carboxyphenoxy propane
PubChem CID 138500
IUPAC Name 4-propoxybenzoic acid
SMILES CCCOC1=CC=C(C=C1)C(O)=O

Specifications

Melting Point 145°C to 147°C
Quantity 5 g
Beilstein 2208387
Formula Weight 180.2
Percent Purity 98%
Chemical Name or Material 4-n-Propoxybenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only