accessibility menu, dialog, popup

UserName

CAS RN 5438-19-7

IUPAC Name: 4-propoxybenzoic acid
Synonyms: 4-propoxybenzoic acid
Molecular Weight (g/mol): 180.2
Molecular Formula: C10H12O3
InChi Key: GDFUWFOCYZZGQU-UHFFFAOYSA-N
SMILES: CCCOC1=CC=C(C=C1)C(O)=O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)