missing translation for 'onlineSavingsMsg'
Learn More

4-n-Pentylbenzaldehyde, 95%

Catalog No. AAL0920806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0920806 5 g
AAL0920803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0920806 Supplier Thermo Scientific Chemicals Supplier No. L0920806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6853-57-2 | C12H16O | 176.259 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6853-57-2
Molecular Formula C12H16O
Molecular Weight (g/mol) 176.259
MDL Number MFCD00043599
InChI Key NQVZPRUSNWNSQH-UHFFFAOYSA-N
Synonym p-pentylbenzaldehyde, benzaldehyde, 4-pentyl, 4-n-pentylbenzaldehyde, p-amylbenzaldehyde, 4-n-amylbenzaldehyde, p-n-pentylbenzaldehyde, p-pentyl-benzaldehyde, p-n-pentylben-zaldehyde, p-pentylbenzaldehyde, i
PubChem CID 23290
IUPAC Name 4-pentylbenzaldehyde
SMILES CCCCCC1=CC=C(C=C1)C=O

Specifications

Boiling Point 142°C to 144°C (2 mmHg)
Refractive Index 1.5215
Quantity 5 g
Beilstein 1869642
Sensitivity Air Sensitive
Formula Weight 176.26
Percent Purity 95%
Chemical Name or Material 4-n-Pentylbenzaldehyde
EINECSNumber 000-000-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only