missing translation for 'onlineSavingsMsg'
Learn More

4'-n-Octyloxybiphenyl-4-carboxylic acid, 99%

CAS: 59748-18-4 | C21H26O3 | 326.44 g/mol

$76.96 - $264.79

Chemical Identifiers

CAS 59748-18-4
Molecular Formula C21H26O3
Molecular Weight (g/mol) 326.44
MDL Number MFCD00192369
InChI Key YNBBQLUKHHSKPW-UHFFFAOYSA-N
Synonym 4'-octyloxy-1,1'-biphenyl-4-carboxylic acid, 4'-octyloxy-4-biphenylcarboxylic acid, 4-n-octyloxybiphenyl-4'-carboxylic acid, 4'-octyloxy biphenyl-4-carboxylic acid, 4-4-octyloxyphenyl benzoic acid, 4-4-octoxyphenyl benzoic acid, 4-4-octyloxy phenyl benzoic acid, 4-octyloxybiphenyl-4'carboxylic acid
PubChem CID 2802556
IUPAC Name 4-(4-octoxyphenyl)benzoic acid
SMILES CCCCCCCCOC1=CC=C(C=C1)C1=CC=C(C=C1)C(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2221603
View Documents
Thermo Scientific Chemicals
B2221603
1 g
Each for $76.96
Only null left
 
AAB2221606
View Documents
Thermo Scientific Chemicals
B2221606
5 g
Each for $264.79
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

59748-18-4
326.44
YNBBQLUKHHSKPW-UHFFFAOYSA-N
2802556
CCCCCCCCOC1=CC=C(C=C1)C1=CC=C(C=C1)C(O)=O
C21H26O3
MFCD00192369
4'-octyloxy-1,1'-biphenyl-4-carboxylic acid, 4'-octyloxy-4-biphenylcarboxylic acid, 4-n-octyloxybiphenyl-4'-carboxylic acid, 4'-octyloxy biphenyl-4-carboxylic acid, 4-4-octyloxyphenyl benzoic acid, 4-4-octoxyphenyl benzoic acid, 4-4-octyloxy phenyl benzoic acid, 4-octyloxybiphenyl-4'carboxylic acid
4-(4-octoxyphenyl)benzoic acid

Specifications

59748-18-4
MFCD00192369
4'-octyloxy-1,1'-biphenyl-4-carboxylic acid, 4'-octyloxy-4-biphenylcarboxylic acid, 4-n-octyloxybiphenyl-4'-carboxylic acid, 4'-octyloxy biphenyl-4-carboxylic acid, 4-4-octyloxyphenyl benzoic acid, 4-4-octoxyphenyl benzoic acid, 4-4-octyloxy phenyl benzoic acid, 4-octyloxybiphenyl-4'carboxylic acid
CCCCCCCCOC1=CC=C(C=C1)C1=CC=C(C=C1)C(O)=O
326.44
326.44
4'-n-Octyloxybiphenyl-4-carboxylic acid
C21H26O3
1 g
YNBBQLUKHHSKPW-UHFFFAOYSA-N
4-(4-octoxyphenyl)benzoic acid
2802556
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only