missing translation for 'onlineSavingsMsg'
Learn More

4-n-Octyloxybenzonitrile, 98%

Catalog No. AAA1284306
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1284306 5 g
AAA1284314 25 g
AAA1284322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1284306 Supplier Thermo Scientific Chemicals Supplier No. A1284306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 88374-55-4 | C15H21NO | 231.339 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 88374-55-4
Molecular Formula C15H21NO
Molecular Weight (g/mol) 231.339
MDL Number MFCD00043483
InChI Key GFNSBTARZPEIPN-UHFFFAOYSA-N
Synonym 4-n-octyloxybenzonitrile, p-octyloxybenzonitrile, 4-octyloxy benzonitrile, 4-octyloxybenzonitrile, p-octyloxy benzonitrile, 4-octyloxybenzenecarbonitrile, 4-n-octyloxycyanobenzene, 4-octyloxy benzonitrile #
PubChem CID 145161
IUPAC Name 4-octoxybenzonitrile
SMILES CCCCCCCCOC1=CC=C(C=C1)C#N

Specifications

Melting Point 24°C to 26°C
Density 0.937
Boiling Point 171°C to 173°C (2 mmHg)
Refractive Index 1.51
Quantity 5 g
UN Number UN3439
Beilstein 2844884
Formula Weight 231.34
Percent Purity 98%
Chemical Name or Material 4-n-Octyloxybenzonitrile
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only