missing translation for 'onlineSavingsMsg'
Learn More

4-n-Nonyloxyaniline, 98%

Catalog No. AAL0912604
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0912604 2 g
AAL0912609 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0912604 Supplier Thermo Scientific Chemicals Supplier No. L0912604
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 50262-67-4 | C15H25NO | 235.371 g/mol

It is employed as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediate for pharmaceutical.

Solubility
Insoluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 50262-67-4
Molecular Formula C15H25NO
Molecular Weight (g/mol) 235.371
MDL Number MFCD00043622
InChI Key JQLBBFVOAHUASD-UHFFFAOYSA-N
Synonym p-nonyloxyaniline, 4-n-nonyloxyaniline, 4-nonyloxyaniline, 4-nonyloxy aniline, 4-nonyloxyphenylamine, 4-nonyloxy aniline #, benzenamine,4-nonyloxy
PubChem CID 142698
IUPAC Name 4-nonoxyaniline
SMILES CCCCCCCCCOC1=CC=C(C=C1)N

Specifications

Melting Point 36°C to 42°C
Boiling Point 176°C to 179°C (2 mmHg)
Quantity 2 g
UN Number UN2811
Beilstein 4232321
Solubility Information Insoluble in water.
Formula Weight 235.37
Percent Purity 98%
Chemical Name or Material 4-n-Nonyloxyaniline
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only