Learn More
4-n-Hexylresorcinol, 99%
CAS: 136-77-6 | C12H18O2 | 194.27 g/mol
Supplier: Thermo Scientific Chemicals A1255214
Description
4-Hexylresorcinol is an inhibitor of transglutaminase-2 in KB cells. Studies indicate that 4-Hexylresorcinol reduces DNA oxidative damage in human lymphocytes. is a substituted dihydroxybenzene used topically as an antiseptic for the treatment of minor skin infections. is a substituted phenol with bactericidal, antihelminthic and potential antineoplastic activities. The combination of 4-Hexylresorcinol and Cisplatin has been shown to inhibit tumor growth, cell growth, and KB cell viability.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Hexylresorcinol is an inhibitor of transglutaminase-2 in KB cells. Studies indicate that 4-Hexylresorcinol reduces DNA oxidative damage in human lymphocytes. is a substituted dihydroxybenzene used topically as an antiseptic for the treatment of minor skin infections. is a substituted phenol with bactericidal, antihelminthic and potential antineoplastic activities. The combination of 4-Hexylresorcinol and Cisplatin has been shown to inhibit tumor growth, cell growth, and KB cell viability.
Solubility
Soluble in water. 0.05 g/100 mL (18°C)
Notes
Incompatible with acid chlorides, acid anhydrides, oxidizing agents. Store at room temperature.
Specifications
| 4-n-Hexylresorcinol | |
| 64°C to 67°C | |
| >100°C (230°F) | |
| MFCD00002284 | |
| 2048312 | |
| hexylresorcinol, 4-hexylresorcinol, antascarin, ascaricid, ascarinol, 4-n-hexylresorcinol, oxana, p-hexylresorcinol, adrover, caprokol | |
| WFJIVOKAWHGMBH-UHFFFAOYSA-N | |
| 4-hexylbenzene-1,3-diol | |
| 3610 | |
| 99% |
| 136-77-6 | |
| 333°C to 335°C | |
| C12H18O2 | |
| 25 g | |
| 14,4712 | |
| Soluble in water. 0.05g/100 mL (18°C) | |
| CCCCCCC1=CC=C(O)C=C1O | |
| 194.27 | |
| 194.27 |
Chemical Identifiers
| 136-77-6 | |
| 194.27 | |
| WFJIVOKAWHGMBH-UHFFFAOYSA-N | |
| 3610 | |
| CCCCCCC1=CC=C(O)C=C1O |
| C12H18O2 | |
| MFCD00002284 | |
| hexylresorcinol, 4-hexylresorcinol, antascarin, ascaricid, ascarinol, 4-n-hexylresorcinol, oxana, p-hexylresorcinol, adrover, caprokol | |
| 4-hexylbenzene-1,3-diol |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 205-257-4
RTECSNumber : VH1575000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only