accessibility menu, dialog, popup

UserName

CAS RN 136-77-6

IUPAC Name: 4-hexylbenzene-1,3-diol
Synonyms: oxana worm-agen cystoids anthelmintic hexylresorcin hexylresorzin ascarinol hexylresorcinolum 己雷琐辛 prensol hexylresorcinol (substance)
Molecular Weight (g/mol): 194.27
Molecular Formula: C12H18O2
InChi Key: WFJIVOKAWHGMBH-UHFFFAOYSA-N
SMILES: CCCCCCC1=CC=C(O)C=C1O
Boiling Point 
  • (1)
  • (3)
  • (2)
Percent Purity 
  • (1)
  • (2)
  • (3)
Quantity 
  • (2)
  • (3)
  • (1)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (5)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)