missing translation for 'onlineSavingsMsg'
Learn More

4-n-Heptylphenol, 98+%

Catalog No. AAA1540703
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1540703 1 g
AAA1540706 5 g
AAA1540714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1540703 Supplier Thermo Scientific Chemicals Supplier No. A1540703
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1987-50-4 | C13H20O | 192.302 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1987-50-4
Molecular Formula C13H20O
Molecular Weight (g/mol) 192.302
MDL Number MFCD00041751
InChI Key KNDDEFBFJLKPFE-UHFFFAOYSA-N
Synonym 4-n-heptylphenol, phenol, 4-heptyl, p-heptylphenol, heptanyl-p-phenol, p-hydroxyheptylbenzene, phenol, heptyl derivs., p-n-heptylphenol, unii-i743z35dvb, phenol, heptenylated, p-heptyl phenol
PubChem CID 16143
ChEBI CHEBI:34438
IUPAC Name 4-heptylphenol
SMILES CCCCCCCC1=CC=C(C=C1)O

Specifications

Melting Point 25°C to 28°C
Boiling Point 156°C (9 mmHg)
Refractive Index 1.507
Quantity 1 g
UN Number UN2430
Formula Weight 192.3
Percent Purity ≥98%
Chemical Name or Material 4-n-Heptylphenol
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ALKYLPHENOLS, SOLID, N.O.S.
EINECSNumber 217-862-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only