accessibility menu, dialog, popup

UserName

CAS RN 1987-50-4

IUPAC Name: 4-heptylphenol
Synonyms: heptanyl-P-phenol p-hydroxyheptylbenzene
Molecular Weight (g/mol): 192.3
Molecular Formula: C13H20O
InChi Key: KNDDEFBFJLKPFE-UHFFFAOYSA-N
SMILES: CCCCCCCC1=CC=C(O)C=C1
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)