missing translation for 'onlineSavingsMsg'
Learn More

4-n-Dodecylbenzoic acid, 98%

Catalog No. AAH3198303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3198303 1 g
AAH3198306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3198303 Supplier Thermo Scientific Chemicals Supplier No. H3198303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 21021-55-6 | C19H30O2 | 290.447 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 21021-55-6
Molecular Formula C19H30O2
Molecular Weight (g/mol) 290.447
MDL Number MFCD04972235
InChI Key JCHCIOJFCCQEKZ-UHFFFAOYSA-N
Synonym p-dodecylbenzoic acid, benzoic acid, 4-dodecyl, p-n-dodecyl benzoic acid, benzoicacid,4-dodecyl, 4-n-dodecylbenzoic acid
PubChem CID 413311
IUPAC Name 4-dodecylbenzoic acid
SMILES CCCCCCCCCCCCC1=CC=C(C=C1)C(=O)O

Specifications

Quantity 1 g
Formula Weight 290.45
Percent Purity 98%
Chemical Name or Material 4-n-Dodecylbenzoic acid
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only