missing translation for 'onlineSavingsMsg'
Learn More

4-n-Decyloxybenzoic acid, 96%

Catalog No. p-7022094
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1470306 5 g
AAA1470314 25 g
AAA1470322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1470306 Supplier Thermo Scientific Chemicals Supplier No. A1470306
Only null left

CAS: 5519-23-3 | C17H26O3 | 278.392 g/mol

4-n-Decyloxybenzoic acid, is used in the syntheis of liquid crystals. It can also be used to react with 3-bromo-propene to produce 5-allyloxy-2-phenyl-[1,3]dioxane. This reaction will need reagent tetra-n-butylammonium bromide and NaH, and the menstruum dimethylformamide. The reaction time is 2 hour with temperature of 20°C,

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-n-Decyloxybenzoic acid, is used in the syntheis of liquid crystals. It can also be used to react with 3-bromo-propene to produce 5-allyloxy-2-phenyl-[1,3]dioxane. This reaction will need reagent tetra-n-butylammonium bromide and NaH, and the menstruum dimethylformamide. The reaction time is 2 hour with temperature of 20°C,

Solubility
Soluble in water.

Notes
m.p. 97°(S), 122°(N), 142°(I)

Chemical Identifiers

CAS 5519-23-3
Molecular Formula C17H26O3
Molecular Weight (g/mol) 278.392
MDL Number MFCD00020360
InChI Key NZNICZRIRMGOFG-UHFFFAOYSA-N
PubChem CID 138527
IUPAC Name 4-decoxybenzoic acid
SMILES CCCCCCCCCCOC1=CC=C(C=C1)C(=O)O

Specifications

Melting Point 97°C (S)
Quantity 5 g
Beilstein 2113730
Synonym 4-decyloxy benzoic acid, p-decyloxybenzoic acid, 4-n-decyloxybenzoic acid, 4-decyloxybenzoic acid, benzoic acid,4-decyloxy, benzoic acid, 4-decyloxy, pubchem2671, p-n-decyloxybenzoic acid, 4-decyloxy benzoicacid, para-decyloxy benzoic acid
Solubility Information Soluble in water.
Formula Weight 278.39
Percent Purity 96%
Chemical Name or Material 4-n-Decyloxybenzoic acid
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only