accessibility menu, dialog, popup

UserName

H3COOHO

CAS RN 5519-23-3

IUPAC Name: 4-(decyloxy)benzoic acid
Synonyms: benzoic acid, 4-(decyloxy)-
Molecular Weight (g/mol): 278.39
Molecular Formula: C17H26O3
InChi Key: NZNICZRIRMGOFG-UHFFFAOYSA-N
SMILES: CCCCCCCCCCOC1=CC=C(C=C1)C(O)=O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)