missing translation for 'onlineSavingsMsg'
Learn More

4-n-Butoxybenzoic acid, 98%

Catalog No. AAB2150614
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2150614 25 g
AAB2150622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2150614 Supplier Thermo Scientific Chemicals Supplier No. B2150614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1498-96-0 | C11H14O3 | 194.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1498-96-0
Molecular Formula C11H14O3
Molecular Weight (g/mol) 194.23
MDL Number MFCD00002546
InChI Key LAUFPZPAKULAGB-UHFFFAOYSA-N
Synonym p-butoxybenzoic acid, 4-n-butoxybenzoic acid, benzoic acid, 4-butoxy, benzoic acid, p-butoxy, p-butyloxybenzoic acid, 4-butoxy-benzoic acid, 4-butyloxybenzoic acid, p-n-butoxybenzoic acid, p-n-butyloxybenzoic acid, 4-n-butyloxybenzoic acid
PubChem CID 72971
IUPAC Name 4-butoxybenzoic acid
SMILES CCCCOC1=CC=C(C=C1)C(=O)O

Specifications

Melting Point 146°C to 147°C (C-N)
Quantity 25 g
Formula Weight 194.23
Percent Purity 98%
Chemical Name or Material 4-n-Butoxybenzoic acid
EINECSNumber 216-104-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only