accessibility menu, dialog, popup

UserName

H3COOHO

CAS RN 1498-96-0

IUPAC Name: 4-butoxybenzoic acid
Synonyms: P-butoxybenzoic acid
Molecular Weight (g/mol): 194.23
Molecular Formula: C11H14O3
InChi Key: LAUFPZPAKULAGB-UHFFFAOYSA-N
SMILES: CCCCOC1=CC=C(C=C1)C(O)=O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)