Learn More
4-Morpholineacetic acid hydrochloride, 95%
CAS: 89531-58-8 | C6H12ClNO3 | 181.62 g/mol
Supplier: Thermo Scientific Chemicals H6118806
Description
4-Morpholineacetic acid hydrochloride is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Morpholineacetic acid hydrochloride is used as a pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
| 4-Morpholineacetic acid hydrochloride | |
| White | |
| MFCD03540888 | |
| 3701710 | |
| Insoluble in water. | |
| [H+].[Cl-].OC(=O)CN1CCOCC1 | |
| 181.62 | |
| 181.62 | |
| Powder |
| 89531-58-8 | |
| C6H12ClNO3 | |
| 5 g | |
| 2-morpholinoacetic acid hydrochloride, 2-morpholin-4-yl acetic acid hydrochloride, morpholin-4-ylacetic acid hydrochloride, morpholin-4-yl-acetic acid hydrochloride, 4-morpholinacetic acid hcl, 4-morpholineacetic acid hydrochloride, 4-morpholineaceticacid, hydrochloride, 4-morpholinylacetic acid hydrochloride, 4-morpholineacetic acid, hydrochloride, 4-morpholineaceticacid, hydrochloride 1:1 | |
| ICGYBJNONRDZOP-UHFFFAOYSA-N | |
| 2-morpholin-4-ylacetic acid;hydrochloride | |
| 17750946 | |
| 95% |
Chemical Identifiers
| 89531-58-8 | |
| 181.62 | |
| ICGYBJNONRDZOP-UHFFFAOYSA-N | |
| 17750946 | |
| [H+].[Cl-].OC(=O)CN1CCOCC1 |
| C6H12ClNO3 | |
| MFCD03540888 | |
| 2-morpholinoacetic acid hydrochloride, 2-morpholin-4-yl acetic acid hydrochloride, morpholin-4-ylacetic acid hydrochloride, morpholin-4-yl-acetic acid hydrochloride, 4-morpholinacetic acid hcl, 4-morpholineacetic acid hydrochloride, 4-morpholineaceticacid, hydrochloride, 4-morpholinylacetic acid hydrochloride, 4-morpholineacetic acid, hydrochloride, 4-morpholineaceticacid, hydrochloride 1:1 | |
| 2-morpholin-4-ylacetic acid;hydrochloride |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only