missing translation for 'onlineSavingsMsg'
Learn More

4-Morpholineacetic acid hydrochloride, 95%

Catalog No. p-7053621
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH61188MD 250 mg
AAH6118803 1 g
AAH6118806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH61188MD Supplier Thermo Scientific Chemicals Supplier No. H61188MD
Only null left

CAS: 89531-58-8 | C6H12ClNO3 | 181.62 g/mol

4-Morpholineacetic acid hydrochloride is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Morpholineacetic acid hydrochloride is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 89531-58-8
Molecular Formula C6H12ClNO3
Molecular Weight (g/mol) 181.62
MDL Number MFCD03540888
InChI Key ICGYBJNONRDZOP-UHFFFAOYSA-N
Synonym 2-morpholinoacetic acid hydrochloride, 2-morpholin-4-yl acetic acid hydrochloride, morpholin-4-ylacetic acid hydrochloride, morpholin-4-yl-acetic acid hydrochloride, 4-morpholinacetic acid hcl, 4-morpholineacetic acid hydrochloride, 4-morpholineaceticacid, hydrochloride, 4-morpholinylacetic acid hydrochloride, 4-morpholineacetic acid, hydrochloride, 4-morpholineaceticacid, hydrochloride 1:1
PubChem CID 17750946
IUPAC Name 2-morpholin-4-ylacetic acid;hydrochloride
SMILES [H+].[Cl-].OC(=O)CN1CCOCC1

Specifications

Color White
Quantity 250 mg
Beilstein 3701710
Solubility Information Insoluble in water.
Formula Weight 181.62
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 4-Morpholineacetic acid hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only