missing translation for 'onlineSavingsMsg'
Learn More

4-Morpholineacetic acid hydrochloride, 95%

CAS: 89531-58-8 | C6H12ClNO3 | 181.62 g/mol

Supplier:  Thermo Scientific Chemicals H6118803

 View more versions of this product

Catalog No. AAH6118803

Only null left
Explore available promotions

Description

Description

4-Morpholineacetic acid hydrochloride is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Morpholineacetic acid hydrochloride is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Specifications

4-Morpholineacetic acid hydrochloride
White
MFCD03540888
3701710
Insoluble in water.
[H+].[Cl-].OC(=O)CN1CCOCC1
181.62
181.62
Powder
89531-58-8
C6H12ClNO3
1 g
2-morpholinoacetic acid hydrochloride, 2-morpholin-4-yl acetic acid hydrochloride, morpholin-4-ylacetic acid hydrochloride, morpholin-4-yl-acetic acid hydrochloride, 4-morpholinacetic acid hcl, 4-morpholineacetic acid hydrochloride, 4-morpholineaceticacid, hydrochloride, 4-morpholinylacetic acid hydrochloride, 4-morpholineacetic acid, hydrochloride, 4-morpholineaceticacid, hydrochloride 1:1
ICGYBJNONRDZOP-UHFFFAOYSA-N
2-morpholin-4-ylacetic acid;hydrochloride
17750946
95%

Chemical Identifiers

89531-58-8
181.62
ICGYBJNONRDZOP-UHFFFAOYSA-N
17750946
[H+].[Cl-].OC(=O)CN1CCOCC1
C6H12ClNO3
MFCD03540888
2-morpholinoacetic acid hydrochloride, 2-morpholin-4-yl acetic acid hydrochloride, morpholin-4-ylacetic acid hydrochloride, morpholin-4-yl-acetic acid hydrochloride, 4-morpholinacetic acid hcl, 4-morpholineacetic acid hydrochloride, 4-morpholineaceticacid, hydrochloride, 4-morpholinylacetic acid hydrochloride, 4-morpholineacetic acid, hydrochloride, 4-morpholineaceticacid, hydrochloride 1:1
2-morpholin-4-ylacetic acid;hydrochloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only