missing translation for 'onlineSavingsMsg'
Learn More

4-Methylumbelliferone 98.0+%, TCI America™

Supplier:  TCI America M0453100G

 View more versions of this product

Catalog No. M0453100G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

4-Methylumbelliferone
188°C
C10H8O3
100 g
HSHNITRMYYLLCV-UHFFFAOYSA-N
7-hydroxy-4-methyl-2H-chromen-2-one
5280567
176.17
Crystalline Powder
90-33-5
White-Yellow
MFCD00006866
4-methylumbelliferone, hymecromone, 7-hydroxy-4-methylcoumarin, imecromone, 7-hydroxy-4-methyl-2h-chromen-2-one, cholestil, mendiaxon, 4-methyl-7-hydroxycoumarin, beta-methylumbelliferone, bilcolic
CC1=CC(=O)OC2=CC(O)=CC=C12
176.17
CHEBI:17224
≥98.0% (T)

Chemical Identifiers

90-33-5
176.17
HSHNITRMYYLLCV-UHFFFAOYSA-N
5280567
7-hydroxy-4-methyl-2H-chromen-2-one
C10H8O3
MFCD00006866
4-methylumbelliferone, hymecromone, 7-hydroxy-4-methylcoumarin, imecromone, 7-hydroxy-4-methyl-2h-chromen-2-one, cholestil, mendiaxon, 4-methyl-7-hydroxycoumarin, beta-methylumbelliferone, bilcolic
CHEBI:17224
CC1=CC(=O)OC2=CC(O)=CC=C12
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (9)-1048

RTECSNumber : GN7000000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions