missing translation for 'onlineSavingsMsg'
Learn More

4-Methylumbelliferone 98.0+%, TCI America™

Catalog No. p-7132181
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M045325G 25 g
M0453100G 100 g
M0453500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M045325G Supplier TCI America Supplier No. M045325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 90-33-5
Molecular Formula C10H8O3
Molecular Weight (g/mol) 176.17
MDL Number MFCD00006866
InChI Key HSHNITRMYYLLCV-UHFFFAOYSA-N
Synonym 4-methylumbelliferone, hymecromone, 7-hydroxy-4-methylcoumarin, imecromone, 7-hydroxy-4-methyl-2h-chromen-2-one, cholestil, mendiaxon, 4-methyl-7-hydroxycoumarin, beta-methylumbelliferone, bilcolic
PubChem CID 5280567
ChEBI CHEBI:17224
IUPAC Name 7-hydroxy-4-methyl-2H-chromen-2-one
SMILES CC1=CC(=O)OC2=CC(O)=CC=C12

Specifications

Melting Point 188°C
Color White-Yellow
Quantity 25 g
Formula Weight 176.17
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methylumbelliferone
EINECSNumber (9)-1048
RTECSNumber GN7000000
TSCA Yes