accessibility menu, dialog, popup

UserName

CH3OOOH

CAS RN 90-33-5

IUPAC Name: 7-hydroxy-4-methyl-2H-chromen-2-one
Synonyms: 4-methylumbelliferone bilcolic eurogale pilot 447 unichol cumarote-C cantabiline sodium bilicante mendiaxon cantabilin
Molecular Weight (g/mol): 176.17
Molecular Formula: C10H8O3
InChi Key: HSHNITRMYYLLCV-UHFFFAOYSA-N
SMILES: CC1=CC(=O)OC2=CC(O)=CC=C12
Percent Purity 
  • (3)
  • (6)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (6)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (6)
  • (3)
  • (6)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (6)
  • (3)