missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylthio)benzyl Alcohol 98.0+%, TCI America™

Catalog No. p-7133155
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
M23145G 5 g
M231425G 25 g
2 options

Catalog No. M23145G

Supplier: TCI America M23145G

Only null left

Chemical Identifiers

CAS 3446-90-0
Molecular Formula C8H10OS
Molecular Weight (g/mol) 154.227
MDL Number MFCD00009706
InChI Key MTXQKSQYMREAGJ-UHFFFAOYSA-N
Synonym 4-methylthio benzyl alcohol, 4-methylthio phenyl methanol, p-methylthiobenzyl alcohol, 4-methylsulfanyl phenyl methanol, 4-methylthiobenzyl alcohol, 4-methylthio benzylalcohol, p-methylthio benzyl alcohol, 4-methylsulfanylphenyl methanol, 4-hydroxymethyl thioanisole
PubChem CID 592968
IUPAC Name (4-methylsulfanylphenyl)methanol
SMILES CSC1=CC=C(C=C1)CO

Specifications

Melting Point 41°C
Color White
Boiling Point 109°C
Quantity 5 g
Formula Weight 154.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4-(Methylthio)benzyl Alcohol