accessibility menu, dialog, popup

UserName

H3CSOH

CAS RN 3446-90-0

IUPAC Name: [4-(methylsulfanyl)phenyl]methanol
Synonyms: [4-(methylsulfanyl)phenyl]methanol
Molecular Weight (g/mol): 154.23
Molecular Formula: C8H10OS
InChi Key: MTXQKSQYMREAGJ-UHFFFAOYSA-N
SMILES: CSC1=CC=C(CO)C=C1
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)