missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylthio)benzyl alcohol, 98%

Catalog No. AAB2245506
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2245506 5 g
AAB2245514 25 g
AAB2245522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2245506 Supplier Thermo Scientific Chemicals Supplier No. B2245506
Only null left

CAS: 3446-90-0 | C8H10OS | 154.227 g/mol

4-(Methylthio)benzenemethanol is used to prepare (4-substituted benzyl)(trifluoromethyl)pyrazoles and -pyrazolones as potent antihyperglycemic agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Methylthio)benzenemethanol is used to prepare (4-substituted benzyl)(trifluoromethyl)pyrazoles and -pyrazolones as potent antihyperglycemic agents.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 3446-90-0
Molecular Formula C8H10OS
Molecular Weight (g/mol) 154.227
MDL Number MFCD00009706
InChI Key MTXQKSQYMREAGJ-UHFFFAOYSA-N
Synonym 4-methylthio benzyl alcohol, 4-methylthio phenyl methanol, p-methylthiobenzyl alcohol, 4-methylsulfanyl phenyl methanol, 4-methylthiobenzyl alcohol, 4-methylthio benzylalcohol, p-methylthio benzyl alcohol, 4-methylsulfanylphenyl methanol, 4-hydroxymethyl thioanisole
PubChem CID 592968
IUPAC Name (4-methylsulfanylphenyl)methanol
SMILES CSC1=CC=C(C=C1)CO

Specifications

Melting Point 41°C to 43°C
Boiling Point 108°C to 109°C (0.4 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
Solubility Information Soluble in water.
Formula Weight 154.23
Percent Purity 98%
Chemical Name or Material 4-(Methylthio)benzyl alcohol
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only