missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Methylthio)benzenethiol 97.0+%, TCI America™
Supplier: TCI America M18235G
Specifications
| 4-(Methylthio)benzenethiol | |
| Yellow | |
| C7H8S2 | |
| 5 g | |
| KYMOWQQIZINTJZ-UHFFFAOYSA-N | |
| 4-methylsulfanylbenzenethiol | |
| 136896 | |
| ≥97.0% (T) |
| 1122-97-0 | |
| 117°C | |
| MFCD00082770 | |
| 4-methylthio thiophenol, 4-methylthio benzenethiol, 4-methylthiothiophenol, 4-methylsulfanyl benzenethiol, 4-methylsulfanyl thiophenol, p-methylthiothiophenol, 4-mercaptothioanisole, pubchem6825, 4-methylthio-thiophenol | |
| CSC1=CC=C(C=C1)S | |
| 156.261 | |
| 156.26 | |
| Liquid |
Chemical Identifiers
| 1122-97-0 | |
| 156.261 | |
| KYMOWQQIZINTJZ-UHFFFAOYSA-N | |
| 136896 | |
| CSC1=CC=C(C=C1)S |
| C7H8S2 | |
| MFCD00082770 | |
| 4-methylthio thiophenol, 4-methylthio benzenethiol, 4-methylthiothiophenol, 4-methylsulfanyl benzenethiol, 4-methylsulfanyl thiophenol, p-methylthiothiophenol, 4-mercaptothioanisole, pubchem6825, 4-methylthio-thiophenol | |
| 4-methylsulfanylbenzenethiol |
Safety and Handling
TSCA : No