missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylthio)aniline, 98%, Thermo Scientific Chemicals

$92.81 - $92.81

Chemical Identifiers

CAS 104-96-1
Molecular Formula C7H9NS
Molecular Weight (g/mol) 139.22
MDL Number MFCD00007889
InChI Key YKFROQCFVXOUPW-UHFFFAOYSA-N
Synonym 4-methylthio aniline, 4-methylmercapto aniline, 4-aminothioanisole, p-thioanisidine, benzenamine, 4-methylthio, p-methylthio aniline, p-aminothioanisole, p-thiomethoxyaniline, 4-methylsulfanyl aniline, 4-methylthio benzenamine
PubChem CID 66038
SMILES CSC1=CC=C(N)C=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC309830050
SDS View Documents
Thermo Scientific Chemicals
309830050
5 g
Each for $92.81
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

104-96-1
139.22
YKFROQCFVXOUPW-UHFFFAOYSA-N
66038
C7H9NS
MFCD00007889
4-methylthio aniline, 4-methylmercapto aniline, 4-aminothioanisole, p-thioanisidine, benzenamine, 4-methylthio, p-methylthio aniline, p-aminothioanisole, p-thiomethoxyaniline, 4-methylsulfanyl aniline, 4-methylthio benzenamine
CSC1=CC=C(N)C=C1

Specifications

104-96-1
100.0
1.1190g/mL
>110°C
97.5% min. (GC)
C7H9NS
CH3SC6H4NH2
5 g
4-methylthio aniline, 4-methylmercapto aniline, 4-aminothioanisole, p-thioanisidine, benzenamine, 4-methylthio, p-methylthio aniline, p-aminothioanisole, p-thiomethoxyaniline, 4-methylsulfanyl aniline, 4-methylthio benzenamine
CSC1=CC=C(N)C=C1
139.22
139.23
Liquid
97.5
Brown-Red to Orange
272.0°C to 273.0°C
Authentic
Glass bottle
1.6380 to 1.6400
MFCD00007889
1.119
YKFROQCFVXOUPW-UHFFFAOYSA-N
4-methylsulfanylaniline
66038
98%
4-(Methylthio)aniline, 98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 203-256-3

RTECSNumber : BY6302000

TSCA : TSCA

Recommended Storage : Product darkens during storage

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only